Anti-P4HB Mouse Monoclonal Antibody [clone: 537331]

Supplier: R & D Systems
MAB4236
102885-700EA 594.37 USD
102885-700
Anti-P4HB Mouse Monoclonal Antibody [clone: 537331]
Antibodies
Type: Primary
Antigen: P4HB
Clonality: Monoclonal
Clone: 537331
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR