Anti-ITGA11 Rat Monoclonal Antibody [clone: 396214]

Supplier: R & D Systems
MAB4235
220020-813EA 600.15 USD
220020-813
Anti-ITGA11 Rat Monoclonal Antibody [clone: 396214]
Antibodies
Type: Primary
Antigen: ITGA11
Clonality: Monoclonal
Clone: 396214
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR