Anti-PPP1R1B Rat Monoclonal Antibody [clone: 375604]

Supplier: R & D Systems
MAB4230
220020-812EA 600.15 USD
220020-812
Anti-PPP1R1B Rat Monoclonal Antibody [clone: 375604]
Antibodies
Type: Primary
Antigen: PPP1R1B
Clonality: Monoclonal
Clone: 375604
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR