Anti-ACKR3 Mouse Monoclonal Antibody [clone: 11G8]

Supplier: R & D Systems
MAB42273
220020-810EA 647.88 USD
220020-810
Anti-ACKR3 Mouse Monoclonal Antibody [clone: 11G8]
Antibodies
Type: Primary
Antigen: ACKR3
Clonality: Monoclonal
Clone: 11G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR