Anti-ACKR3 Mouse Monoclonal Antibody [clone: 358426]

Supplier: R & D Systems
MAB4227
220020-809EA 594.37 USD
220020-809
Anti-ACKR3 Mouse Monoclonal Antibody [clone: 358426]
Antibodies
Type: Primary
Antigen: ACKR3
Clonality: Monoclonal
Clone: 358426
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR