Anti-MS4A1 Mouse Monoclonal Antibody [clone: 396444]

Supplier: R & D Systems
MAB4225
220020-808EA 594.37 USD
220020-808
Anti-MS4A1 Mouse Monoclonal Antibody [clone: 396444]
Antibodies
Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: 396444
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR