Anti-CLDN4 Mouse Monoclonal Antibody [clone: 382321]

Supplier: R & D Systems
MAB4219
220020-804EA 639.2 USD
220020-804
Anti-CLDN4 Mouse Monoclonal Antibody [clone: 382321]
Antibodies
Type: Primary
Antigen: CLDN4
Clonality: Monoclonal
Clone: 382321
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR