Anti-SCGB1A1 Rat Monoclonal Antibody [clone: 394324]

Supplier: R & D Systems
MAB4218
220020-803EA 623.29 USD
220020-803
Anti-SCGB1A1 Rat Monoclonal Antibody [clone: 394324]
Antibodies
Type: Primary
Antigen: SCGB1A1
Clonality: Monoclonal
Clone: 394324
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR