Anti-DVL2 Mouse Monoclonal Antibody [clone: 373413]

Supplier: R & D Systems
MAB4216
220020-802EA 577.02 USD
220020-802
Anti-DVL2 Mouse Monoclonal Antibody [clone: 373413]
Antibodies
Type: Primary
Antigen: DVL2
Clonality: Monoclonal
Clone: 373413
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR