Anti-EXTL1 Rat Monoclonal Antibody [clone: 353713]

Supplier: R & D Systems
MAB4214
220020-800EA 561.11 USD
220020-800
Anti-EXTL1 Rat Monoclonal Antibody [clone: 353713]
Antibodies
Type: Primary
Antigen: EXTL1
Clonality: Monoclonal
Clone: 353713
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR