Anti-ALK Mouse Monoclonal Antibody [clone: 383625]

Supplier: R & D Systems
MAB4210
220020-799EA 577.02 USD
220020-799
Anti-ALK Mouse Monoclonal Antibody [clone: 383625]
Antibodies
Type: Primary
Antigen: ALK
Clonality: Monoclonal
Clone: 383625
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR