Anti-CXCL17 Mouse Monoclonal Antibody [clone: 422208]

Supplier: R & D Systems
MAB4207
220020-797EA 594.37 USD
220020-797
Anti-CXCL17 Mouse Monoclonal Antibody [clone: 422208]
Antibodies
Type: Primary
Antigen: CXCL17
Clonality: Monoclonal
Clone: 422208
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR