Anti-MATN4 Mouse Monoclonal Antibody [clone: 401127]

Supplier: R & D Systems
MAB4205
220020-796EA 561.11 USD
220020-796
Anti-MATN4 Mouse Monoclonal Antibody [clone: 401127]
Antibodies
Type: Primary
Antigen: MATN4
Clonality: Monoclonal
Clone: 401127
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR