Anti-ESAM Mouse Monoclonal Antibody [clone: 408519]

Supplier: R & D Systems
MAB4204
220020-794EA 594.37 USD
220020-794
Anti-ESAM Mouse Monoclonal Antibody [clone: 408519]
Antibodies
Type: Primary
Antigen: ESAM
Clonality: Monoclonal
Clone: 408519
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR