Anti-HSPB6 Mouse Monoclonal Antibody [clone: 494310]

Supplier: R & D Systems
MAB4200
220020-793EA 594.37 USD
220020-793
Anti-HSPB6 Mouse Monoclonal Antibody [clone: 494310]
Antibodies
Type: Primary
Antigen: HSPB6
Clonality: Monoclonal
Clone: 494310
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR