Anti-PTGS2 Mouse Monoclonal Antibody [clone: 495222]

Supplier: R & D Systems
MAB4198
220020-791EA 639.2 USD
220020-791
Anti-PTGS2 Mouse Monoclonal Antibody [clone: 495222]
Antibodies
Type: Primary
Antigen: PTGS2
Clonality: Monoclonal
Clone: 495222
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR