Anti-IL11 Rat Monoclonal Antibody [clone: 188507]

Supplier: R & D Systems
MAB4181
220020-788EA 772.25 USD
220020-788
Anti-IL11 Rat Monoclonal Antibody [clone: 188507]
Antibodies
Type: Primary
Antigen: IL11
Clonality: Monoclonal
Clone: 188507
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR