Anti-IL11 Rat Monoclonal Antibody [clone: 188520]

Supplier: R & D Systems
MAB418
220020-787EA 779.48 USD
220020-787
Anti-IL11 Rat Monoclonal Antibody [clone: 188520]
Antibodies
Type: Primary
Antigen: IL11
Clonality: Monoclonal
Clone: 188520
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR