Anti-CAMK2D Mouse Monoclonal Antibody [clone: 438422]

Supplier: R & D Systems
MAB4176
220020-785EA 594.37 USD
220020-785
Anti-CAMK2D Mouse Monoclonal Antibody [clone: 438422]
Antibodies
Type: Primary
Antigen: CAMK2D
Clonality: Monoclonal
Clone: 438422
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR