Anti-PKAC (pan specific) Mouse Monoclonal Antibody [clone: 485013]

Supplier: R & D Systems
MAB4175
220020-784EA 577.02 USD
220020-784
Anti-PKAC (pan specific) Mouse Monoclonal Antibody [clone: 485013]
Antibodies
Type: Primary
Antigen Symbol: PKAC (pan specific)
Antigen Name:
Antigen Synonyms:
Clonality: Monoclonal
Clone: 485013
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR