Anti-CGA Mouse Monoclonal Antibody [clone: 381012]

Supplier: R & D Systems
MAB4169
220020-782EA 594.37 USD
220020-782
Anti-CGA Mouse Monoclonal Antibody [clone: 381012]
Antibodies
Type: Primary
Antigen: CGA
Clonality: Monoclonal
Clone: 381012
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR