Anti-LECT1 Mouse Monoclonal Antibody [clone: 394002]

Supplier: R & D Systems
MAB41681
220020-781EA 577.02 USD
220020-781
Anti-LECT1 Mouse Monoclonal Antibody [clone: 394002]
Antibodies
Type: Primary
Antigen: LECT1
Clonality: Monoclonal
Clone: 394002
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR