Anti-CEACAM3 Mouse Monoclonal Antibody [clone: 417009]

Supplier: R & D Systems
MAB4166
220020-780EA 577.02 USD
220020-780
Anti-CEACAM3 Mouse Monoclonal Antibody [clone: 417009]
Antibodies
Type: Primary
Antigen: CEACAM3
Clonality: Monoclonal
Clone: 417009
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR