Anti-COL23A1 Mouse Monoclonal Antibody [clone: 468642]

Supplier: R & D Systems
MAB4165
102885-694EA 594.37 USD
102885-694
Anti-COL23A1 Mouse Monoclonal Antibody [clone: 468642]
Antibodies
Type: Primary
Antigen: COL23A1
Clonality: Monoclonal
Clone: 468642
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR