Anti-CADM4 Rat Monoclonal Antibody [clone: 404617]

Supplier: R & D Systems
MAB41641
220020-779EA 577.02 USD
220020-779
Anti-CADM4 Rat Monoclonal Antibody [clone: 404617]
Antibodies
Type: Primary
Antigen: CADM4
Clonality: Monoclonal
Clone: 404617
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR