Anti-CADM4 Rat Monoclonal Antibody [clone: 404619]

Supplier: R & D Systems
MAB4164
220020-778EA 561.11 USD
220020-778
Anti-CADM4 Rat Monoclonal Antibody [clone: 404619]
Antibodies
Type: Primary
Antigen: CADM4
Clonality: Monoclonal
Clone: 404619
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR