Anti-CSF1 Rat Monoclonal Antibody [clone: 131614]

Supplier: R & D Systems
MAB4161
220020-777EA 772.25 USD
220020-777
Anti-CSF1 Rat Monoclonal Antibody [clone: 131614]
Antibodies
Type: Primary
Antigen: CSF1
Clonality: Monoclonal
Clone: 131614
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR