Anti-PLXND1 Mouse Monoclonal Antibody [clone: 752815]

Supplier: R & D Systems
MAB41601
102885-692EA 594.37 USD
102885-692
Anti-PLXND1 Mouse Monoclonal Antibody [clone: 752815]
Antibodies
Type: Primary
Antigen: PLXND1
Clonality: Monoclonal
Clone: 752815
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR