Anti-ITGB6 Mouse Monoclonal Antibody [clone: 437216]

Supplier: R & D Systems
MAB41551
220020-775EA 594.37 USD
220020-775
Anti-ITGB6 Mouse Monoclonal Antibody [clone: 437216]
Antibodies
Type: Primary
Antigen: ITGB6
Clonality: Monoclonal
Clone: 437216
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR