Anti-IL1A Mouse Monoclonal Antibody [clone: 409405]

Supplier: R & D Systems
MAB4154
220020-773EA 594.37 USD
220020-773
Anti-IL1A Mouse Monoclonal Antibody [clone: 409405]
Antibodies
Type: Primary
Antigen: IL1A
Clonality: Monoclonal
Clone: 409405
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR