Anti-PPM1A Mouse Monoclonal Antibody [clone: 498922]

Supplier: R & D Systems
MAB4150
220020-772EA 577.02 USD
220020-772
Anti-PPM1A Mouse Monoclonal Antibody [clone: 498922]
Antibodies
Type: Primary
Antigen: PPM1A
Clonality: Monoclonal
Clone: 498922
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR