Anti-HSPA8 Mouse Monoclonal Antibody [clone: 521503]

Supplier: R & D Systems
MAB4148
220020-770EA 594.37 USD
220020-770
Anti-HSPA8 Mouse Monoclonal Antibody [clone: 521503]
Antibodies
Type: Primary
Antigen: HSPA8
Clonality: Monoclonal
Clone: 521503
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR