Anti-BNIP3 Mouse Monoclonal Antibody [clone: 670621]

Supplier: R & D Systems
MAB4147
102885-686EA 577.02 USD
102885-686
Anti-BNIP3 Mouse Monoclonal Antibody [clone: 670621]
Antibodies
Type: Primary
Antigen: BNIP3
Clonality: Monoclonal
Clone: 670621
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR