Anti-ANXA4 Mouse Monoclonal Antibody [clone: 694624]

Supplier: R & D Systems
MAB4146
102885-684EA 594.37 USD
102885-684
Anti-ANXA4 Mouse Monoclonal Antibody [clone: 694624]
Antibodies
Type: Primary
Antigen: ANXA4
Clonality: Monoclonal
Clone: 694624
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR