Anti-DNAJB1 Mouse Monoclonal Antibody [clone: 419401]

Supplier: R & D Systems
MAB4145
220020-769EA 594.37 USD
220020-769
Anti-DNAJB1 Mouse Monoclonal Antibody [clone: 419401]
Antibodies
Type: Primary
Antigen: DNAJB1
Clonality: Monoclonal
Clone: 419401
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR