Anti-APOE Rat Monoclonal Antibody [clone: 395004]

Supplier: R & D Systems
MAB41441
220020-768EA 600.15 USD
220020-768
Anti-APOE Rat Monoclonal Antibody [clone: 395004]
Antibodies
Type: Primary
Antigen: APOE
Clonality: Monoclonal
Clone: 395004
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR