Anti-ACKR1 Mouse Monoclonal Antibody [clone: 35837]

Supplier: R & D Systems
MAB4139
220020-766EA 594.37 USD
220020-766
Anti-ACKR1 Mouse Monoclonal Antibody [clone: 35837]
Antibodies
Type: Primary
Antigen: ACKR1
Clonality: Monoclonal
Clone: 35837
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR