Anti-S100 A4 Rat Monoclonal Antibody [clone: 438709]

Supplier: R & D Systems
MAB4138
220020-765EA 577.02 USD
220020-765
Anti-S100 A4 Rat Monoclonal Antibody [clone: 438709]
Antibodies
Type: Primary
Antigen: S100 A4
Clonality: Monoclonal
Clone: 438709
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR