Anti-DMP1 Mouse Monoclonal Antibody [clone: 560716]

Supplier: R & D Systems
MAB4129
102885-682EA 763.57 USD
102885-682
Anti-DMP1 Mouse Monoclonal Antibody [clone: 560716]
Antibodies
Type: Primary
Antigen: DMP1
Clonality: Monoclonal
Clone: 560716
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR