Anti-CEACAM5 Mouse Monoclonal Antibody [clone: 487609]

Supplier: R & D Systems
MAB41281
220020-763EA 655.11 USD
220020-763
Anti-CEACAM5 Mouse Monoclonal Antibody [clone: 487609]
Antibodies
Type: Primary
Antigen: CEACAM5
Clonality: Monoclonal
Clone: 487609
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR