Anti-APOB Mouse Monoclonal Antibody [clone: 369717]

Supplier: R & D Systems
MAB4124
220020-761EA 639.2 USD
220020-761
Anti-APOB Mouse Monoclonal Antibody [clone: 369717]
Antibodies
Type: Primary
Antigen: APOB
Clonality: Monoclonal
Clone: 369717
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR