Anti-RSPO3 Rat Monoclonal Antibody [clone: 400403]

Supplier: R & D Systems
MAB41201
220020-760EA 600.15 USD
220020-760
Anti-RSPO3 Rat Monoclonal Antibody [clone: 400403]
Antibodies
Type: Primary
Antigen: RSPO3
Clonality: Monoclonal
Clone: 400403
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR