Anti-RSPO3 Rat Monoclonal Antibody [clone: 400404]

Supplier: R & D Systems
MAB4120
220020-759EA 577.02 USD
220020-759
Anti-RSPO3 Rat Monoclonal Antibody [clone: 400404]
Antibodies
Type: Primary
Antigen: RSPO3
Clonality: Monoclonal
Clone: 400404
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR