Anti-IDUA Mouse Monoclonal Antibody [clone: 452619]

Supplier: R & D Systems
MAB4119
220020-758EA 655.11 USD
220020-758
Anti-IDUA Mouse Monoclonal Antibody [clone: 452619]
Antibodies
Type: Primary
Antigen: IDUA
Clonality: Monoclonal
Clone: 452619
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR