Anti-ITGA2B Rat Monoclonal Antibody [clone: 386627]

Supplier: R & D Systems
MAB4118
220020-756EA 594.37 USD
220020-756
Anti-ITGA2B Rat Monoclonal Antibody [clone: 386627]
Antibodies
Type: Primary
Antigen: ITGA2B
Clonality: Monoclonal
Clone: 386627
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR