Anti-RSPO4 Rat Monoclonal Antibody [clone: 435203]

Supplier: R & D Systems
MAB4106
220020-754EA 561.11 USD
220020-754
Anti-RSPO4 Rat Monoclonal Antibody [clone: 435203]
Antibodies
Type: Primary
Antigen: RSPO4
Clonality: Monoclonal
Clone: 435203
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR