Anti-KLK2 Mouse Monoclonal Antibody [clone: 426723]

Supplier: R & D Systems
MAB4104
220020-751EA 616.06 USD
220020-751
Anti-KLK2 Mouse Monoclonal Antibody [clone: 426723]
Antibodies
Type: Primary
Antigen: KLK2
Clonality: Monoclonal
Clone: 426723
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR