Anti-CMA1 Mouse Monoclonal Antibody [clone: 422818]

Supplier: R & D Systems
MAB4099
220020-747EA 594.37 USD
220020-747
Anti-CMA1 Mouse Monoclonal Antibody [clone: 422818]
Antibodies
Type: Primary
Antigen: CMA1
Clonality: Monoclonal
Clone: 422818
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR