Anti-BACE2 Rat Monoclonal Antibody [clone: 391017]

Supplier: R & D Systems
MAB4097
220020-746EA 577.02 USD
220020-746
Anti-BACE2 Rat Monoclonal Antibody [clone: 391017]
Antibodies
Type: Primary
Antigen: BACE2
Clonality: Monoclonal
Clone: 391017
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR