Anti-FKBP4 Rat Monoclonal Antibody [clone: 425123]

Supplier: R & D Systems
MAB4095
220020-745EA 577.02 USD
220020-745
Anti-FKBP4 Rat Monoclonal Antibody [clone: 425123]
Antibodies
Type: Primary
Antigen Symbol: FKBP4
Antigen Name: FK506 binding protein 4, 59kDa
Antigen Synonyms: FKBP51, FKBP52, FKBP59, HBI, Hsp56, PPIase, p52
Clonality: Monoclonal
Clone: 425123
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR